missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium borohydride, 95%, Thermo Scientific™

Product Code. 10556353 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10556353

Brand: Thermo Scientific Acros 376370250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18117-29-8
Molecular Formula C25H22P·BH4
Molecular Weight (g/mol) 368.27
InChI Key ICVMMQSETPKHRH-UHFFFAOYSA-N
Synonym benzyl triphenyl phosphanium;boron 1-, benzyltriphenylphosphonium tetrahydroborate
PubChem CID 11303127
IUPAC Name benzyl(triphenyl)phosphanium;boron(1-)
SMILES [B-].C1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 157.0°C to 163.0°C
Assay Percent Range 95%
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,dichloromethane,insoluble in benzene and hexane
Formula Weight 368.27
Percent Purity 95%
Chemical Name or Material Benzyltriphenylphosphonium borohydride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyltriphenylphosphonium borohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.