accessibility menu, dialog, popup

UserName

CAS RN 18117-29-8

BP

CAS RN 18117-29-8

IUPAC Name: benzyltriphenylphosphanium λ¹-boranuidylidene
Synonyms: benzyltriphenylphosphanium λ¹-boranuidylidene
Molecular Weight (g/mol): 364.23
Molecular Formula: C25H22BP
InChi Key: ICVMMQSETPKHRH-UHFFFAOYSA-N
SMILES: [B-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results