missing translation for 'onlineSavingsMsg'
Learn More

Benzyl butyrate, 98%, Thermo Scientific™

Product Code. 11451287
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11451287

Brand: Thermo Scientific Alfa Aesar B24241.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-37-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00027133
InChI Key VONGZNXBKCOUHB-UHFFFAOYSA-N
Synonym benzyl butyrate, benzyl n-butyrate, butanoic acid, phenylmethyl ester, phenylmethyl butanoate, benzyl n-butanoate, butyric acid, benzyl ester, phenylmethyl butyrate, benzylester kyseliny maselne, benzylbutyrate, benzyl butyrate natural
PubChem CID 7650
IUPAC Name benzyl butanoate
SMILES CCCC(=O)OCC1=CC=CC=C1

Specifications

Density 1.009
Boiling Point 240°C
Flash Point 107°C (224°F)
Refractive Index 1.493
Quantity 250 g
Beilstein 2047625
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material Benzyl butyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.