accessibility menu, dialog, popup

UserName

CAS RN 103-37-7

H3COO

CAS RN 103-37-7

IUPAC Name: benzyl butanoate
Synonyms: benzyl butyrate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: VONGZNXBKCOUHB-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results