Learn More
Benzydamine hydrochloride, Thermo Scientific™
£37.30 - £123.00
Chemical Identifiers
| CAS | 132-69-4 |
|---|---|
| Molecular Formula | C19H24ClN3O |
| Molecular Weight (g/mol) | 345.871 |
| MDL Number | MFCD00078957 |
| InChI Key | HNNIWKQLJSNAEQ-UHFFFAOYSA-N |
| Synonym | benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin |
| PubChem CID | 65464 |
| IUPAC Name | 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride |
| SMILES | CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15423809
|
Thermo Scientific Alfa Aesar
J61180.06 |
5g |
£37.30
5g |
Estimated Shipment: 16-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15433809
|
Thermo Scientific Alfa Aesar
J61180.14 |
25g |
£123.00
25g |
Estimated Shipment: 29-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Nonsteroidal analgesic and anti-inflammatory compound
- A prostaglandin synthase inhibitor
Chemical Identifiers
| C19H24ClN3O | |
| MFCD00078957 | |
| benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin | |
| 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride |
Specifications
| 132-69-4 | |
| MFCD00078957 | |
| benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin | |
| HNNIWKQLJSNAEQ-UHFFFAOYSA-N | |
| 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride | |
| 65464 | |
| Benzydamine hydrochloride |
| C19H24ClN3O | |
| 14,1122 | |
| Soluble in water,ethanol,chloroform,n-butanol or DMSO /n | |
| CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl | |
| 345.871 | |
| 345.87 |
Safety and Handling
GHS H Statement
H302-H312-H332-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
H302+H312+H332-H319
missing translation for 'einecsNumber' : 205-076-0
missing translation for 'rtecsNumber' : NK7875000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- Benzydamine hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contain: Benzydamine hydrochloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.