missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine hydrochloride

Product Code. 15423809
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423809 5 g 5g
15433809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423809 Supplier Thermo Scientific Alfa Aesar Supplier No. J61180.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 132-69-4 | C19H24ClN3O | 345.871 g/mol

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-69-4
Molecular Formula C19H24ClN3O
Molecular Weight (g/mol) 345.871
MDL Number MFCD00078957
InChI Key HNNIWKQLJSNAEQ-UHFFFAOYSA-N
Synonym benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
PubChem CID 65464
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl

Specifications

Melting Point 156°C to 161°C
Color White
Quantity 5 g
Merck Index 14,1122
Solubility Information Soluble in water,ethanol,chloroform,n-butanol or DMSO /n
Formula Weight 345.87
Physical Form Crystalline Powder
Chemical Name or Material Benzydamine hydrochloride
compliance-icons
Product Identifier
  • Benzydamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Benzydamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.