missing translation for 'onlineSavingsMsg'
Learn More

Benzotropine methanesulfonate, Thermo Scientific™

Muscarinic receptor antagonist

£14.71 - £20.50

Chemical Identifiers

CAS 132-17-2
Molecular Formula C22H29NO4S
Molecular Weight (g/mol) 403.54
MDL Number MFCD00074784
InChI Key CPFJLLXFNPCTDW-STYNFMPRSA-N
Synonym benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
PubChem CID 3246155
IUPAC Name (1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
SMILES CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484429
SDS View Documents
Thermo Scientific Alfa Aesar
J61954.MC
100mg
ONLINE EXCLUSIVE PRICE
£15.00
£14.71 / 100mg
Save £0.29
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15464429
SDS View Documents
Thermo Scientific Alfa Aesar
J61954.03
ONLINE EXCLUSIVE PRICE
£21.00
£20.50 / 1g
Save £0.50
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water (81 mg/ml at 25°C), DMSO (50 mg/ml at 25°C), ethanol (81 mg/ml at 25°C), and ether (very slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
Specifications

Chemical Identifiers

132-17-2
403.54
CPFJLLXFNPCTDW-STYNFMPRSA-N
3246155
CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1
C22H29NO4S
MFCD00074784
benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
(1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid

Specifications

132-17-2
MFCD00074784
14,1121
Soluble in water (81mg/ml at 25°C),DMSO (50mg/ml at 25°C),ethanol (81mg/ml at 25°C),and ether (very slightly).
CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1
403.54
403.54
C22H29NO4S
UN2811
benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
CPFJLLXFNPCTDW-STYNFMPRSA-N
(1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
3246155
Benzotropine methanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c

H301+H311-H330

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 205-048-8

missing translation for 'rtecsNumber' : YM3150000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.