missing translation for 'onlineSavingsMsg'
Learn More

Benzotropine methanesulfonate

Product Code. 15484429
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484429 100 mg 100mg
15464429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484429 Supplier Thermo Scientific Alfa Aesar Supplier No. J61954.MC
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Muscarinic receptor antagonist | CAS: 132-17-2 | C22H29NO4S | 403.54 g/mol

Benzotropine methanesulfonate is used to treat the symptoms of Parkinson?s disease and is currently in clincial trials for chronic back pain. It shows anti-muscarinic effects comparable to Atropine, and also demonstrates competitive anti-nicotinic action at the nAChR (nicotinic acetylcholine receptor).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (81 mg/ml at 25°C), DMSO (50 mg/ml at 25°C), ethanol (81 mg/ml at 25°C), and ether (very slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-17-2
Molecular Formula C22H29NO4S
Molecular Weight (g/mol) 403.54
MDL Number MFCD00074784
InChI Key CPFJLLXFNPCTDW-STYNFMPRSA-N
Synonym benztropine mesylate, cogentin, benztropine mesilate, benztropine methanesulfonate, unii-wmj8tl7510, benztropine methylsulfonate, benzatropine mesilate, benzatropine mesylate, benzotropine mesylate, cobrentin methanesulfonate
PubChem CID 3246155
IUPAC Name (1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
SMILES CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C
Color White
Quantity 100 mg
UN Number UN2811
Merck Index 14,1121
Solubility Information Soluble in water (81mg/ml at 25°C),DMSO (50mg/ml at 25°C),ethanol (81mg/ml at 25°C),and ether (very slightly).
Formula Weight 403.54
Physical Form Solid
Chemical Name or Material Benzotropine methanesulfonate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.