missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-6-carboxamide, 96%, Thermo Scientific™

Product Code. 15417157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417157 1 g 1g
15427157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15417157 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H34216.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 101084-51-9
Molecular Formula C8H6N2OS
Molecular Weight (g/mol) 178.209
MDL Number MFCD11643189
InChI Key IQFCJGFQUZORLM-UHFFFAOYSA-N
Synonym 6-benzothiazolecarboxamide, benzothiazole-6-carboxamide, benzo d thiazole-6-carboxamide, acmc-20m45a, 6-benzo d thiazolecarboxamide, 6-benzothiazolecarboxamide 6ci
PubChem CID 43236343
IUPAC Name 1,3-benzothiazole-6-carboxamide
SMILES C1=CC2=C(C=C1C(=O)N)SC=N2

Specifikationer

Melting Point 245°C to 246°C
Quantity 1 g
Formula Weight 178.21
Percent Purity 96%
Chemical Name or Material Benzothiazole-6-carboxamide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.