accessibility menu, dialog, popup

UserName

CAS RN 101084-51-9

ONH2SN

CAS RN 101084-51-9

IUPAC Name: 1,3-benzothiazole-6-carboxamide
Synonyms: 1,3-benzothiazole-6-carboxamide
Molecular Weight (g/mol): 178.21
Molecular Formula: C8H6N2OS
InChi Key: IQFCJGFQUZORLM-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C2N=CSC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results