missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,3-dithiocarboxamide, 97%, Thermo Scientific™

Product Code. 15498327
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15498327

Brand: Thermo Scientific Alfa Aesar H51853.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3030-54-4
Molecular Formula C8H8N2S2
Molecular Weight (g/mol) 196.286
MDL Number MFCD16293899
InChI Key DJOXZAYSEDLTGM-UHFFFAOYSA-N
Synonym benzene-1,3-dithiocarboxamide, 1,3-benzenedicarbothioamide, dithioisophthalamid, dithioisophthalamide, 1,3-benzenebis carbothioamide, benzene-1,3-bis carbothioamide
PubChem CID 3896847
IUPAC Name benzene-1,3-dicarbothioamide
SMILES C1=CC(=CC(=C1)C(=S)N)C(=S)N

Specifications

Melting Point 198°C to 200°C
Quantity 5 g
UN Number UN2811
Beilstein 2613862
Formula Weight 196.28
Percent Purity 97%
Chemical Name or Material Benzene-1,3-dithiocarboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.