accessibility menu, dialog, popup

UserName

CAS RN 3030-54-4

NH2SNH2S

CAS RN 3030-54-4

IUPAC Name: benzene-1,3-dicarbothioamide
Synonyms: benzene-1,3-dicarbothioamide
Molecular Weight (g/mol): 196.29
Molecular Formula: C8H8N2S2
InChi Key: DJOXZAYSEDLTGM-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=CC=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results