missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
Description
- Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
- Flat-bottom shape permits use as a weighing boat
- For one-time use
Order Info
100 individually sterile wrapped spoons or 12 nonsterile spoons per pack
Specifications
Specifications
| Quantity | 12/Pk. |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Shape | Flat Bottom |
| Capacity (Metric) | 5 mL |
| Color | White |
| Product Type | Spoon |
| For Use With (Application) | Permits quick sampling of specific sizes of dry or liquid measurements and also provides durability and stiffness when sampling packed powders |
| Material | Polystyrene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?