missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Earth Friendly Wide Handle Sampling Spoons

Lessen environmental impact with PLA (polylactic acid) resin sampling spoons

Brand:  Bel-Art™ F36740-1025

Code : 04

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13492479

  • £12.10 / Pack of 10
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.

  • FDA grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
  • Optimized for retrieving small, specific volume samples of powders or liquids
  • Wide handles allows user to grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials
  • Flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
  • Nonsterile

Specifications

Specifications

Earth Friendly Sampling Spoon
Flat
20 mL
FDA
PLA Resin
Wide
Non-sterile
Designed for retrieving small, specific volume samples of powders or liquids, used as a green alternative to petroleum-based sampling tools
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers