missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars

To agitate liquids in round bottom vessels where other stirring bars cannot be used.

Brand:  Bel-Art™ F37126-0012

19.98 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11398713

  • £21.15 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Made from PTFE
  • Fins on top and bottom ensure complete agitation
  • Does not wobble, maintains position in vessel
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclavable
Specifications

Specifications

12.7 mm
12.7mm
0.5 in.
0.5 in.
Stir Bar
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers