missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Plastic Beaker Set, 10 x 100mL

Product Code. 12903267
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12903267

Brand: Hanna Instruments™ HI740036

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for measuring (up to 100mL) or for sample analysis. Hanna Instruments™ Plastic Beaker Set, 10 x 100mL is a set of 10 graduated plastic beakers that are supplied with some Hanna test kits, photometers and meters.

  • Durable plastic
  • Graduated volume markings
  • 10 piece set of 100mL beakers

Specifications

Color Clear
Capacity (Metric) 100 mL
Material Plastic
Quantity 10
Graduated Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.