missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bead Mill 24 Homogenizer PROMO

Product Code. 16328317 Shop All Fisherbrand Products
Change view
Click to view available options
:
Bead Mill 24 Homogenizer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16328317 Bead Mill 24 Homogenizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16328317

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for variety of applications that require grinding, lysing, and homogenization of biological samples prior to molecular extraction. Used with Fisherbrand™ Bead Kits.

Supplied with FREE pack of 50 x 2 mL tubes prefilled with 2.8 mm ceramic beads, ideal for processing brain, liver, muscle, skin, leaves. RNase/DNase free. Processing range: 300 μL to 1.8 mL

  • Simultaneously homogenize up to 24 x 2 mL or 0.5 mL tubes
  • More samples can be processed in smaller amount of time
  • Convenient front-loading tube holder position for ease of use and optimal bead/sample interactions
  • Processing range is ideal for soft tissue samples
  • No cool-down required between runs (non-stop processing)
  • Disposable tubes ensure no threat of cross-contamination
  • No sample degradation due to quick processing
  • Smaller footprint to maximize bench space
  • Run Time: 1 to 99 sec.
  • Performance Range: 0.8 m/s to 6 m/s in increments of 1.5 m/s
  • Number of Cycles: 1-9

Compliance

CE compliant

TRUSTED_SUSTAINABILITY

Specifications

Product Type Homogenizer
For Use With (Application) DNA/RNA extraction, tissue homogenization, protein purification
Weight (English) 45 lb.
Weight (Metric) 24 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.