missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Antibody (3D9C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £453.00
Specifications
| Antigen | BDNF |
|---|---|
| Clone | 3D9C5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662906
|
Novus Biologicals
NBP2-37276-0.025ml |
0.025 mL |
£197.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18785833
|
Novus Biologicals
NBP2-37276 |
0.1 mL |
£453.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| BDNF | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
| BDNF | |
| IgG1 | |
| Affinity Purified |
| 3D9C5 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Cytokine Research, Immunology, Neuroscience | |
| PBS with 0.05% Sodium Azide | |
| 627 | |
| Purified recombinant fragment of human BDNF (AA: 19-248) expressed in E. Coli. | |
| Primary | |
| 27.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title