missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDNF Antibody (3D9C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37276
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BDNF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
| Mouse | |
| 27.8 kDa | |
| 0.1 mL | |
| Cytokine Research, Immunology, Neuroscience | |
| 627 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3D9C5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BDNF | |
| Purified recombinant fragment of human BDNF (AA: 19-248) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction