missing translation for 'onlineSavingsMsg'
Learn More

BD Prepared Bottled Media: Tryptic Soy Broth

Product Code. 13456526
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13456526 20 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13456526 Supplier BD Supplier No. 257107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5°C to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Tryptic Soy Broth
Packaging Type Bottle
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.