Learn More
BD Phospholipid-binding Protein, APC Annexin V
Has high affinity for PS, and binds to cells with exposed PS
Brand: BD 550474
Additional Details : Weight : 0.01000kg
Beschreibung
- Conjugated to APC
- Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry
Spezifikation
100 Tests | |
Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide | |
Flow Cytometry | |
Liquid |
5 μL | |
4°C | |
APC (Allophycocyanin) |