missing translation for 'onlineSavingsMsg'
Learn More

BD DRAQ5™

Product Code. 15893949 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15893949 200 μL Each
15803959 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15893949 Supplier BD Supplier No. 564902
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to stain nucleic acids in live or fixed cells. BD Pharmingen™ DRAQ5™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or enucleated cells.

  • Far-red fluorescent DNA dye
  • Cell permeable
  • Used to stain nucleic acids in live or fixed cells
  • Has an excitation wavelength maxima of 598/646nm
TRUSTED_SUSTAINABILITY

Specifications

Color Far-red
Concentration 5 mM
Content And Storage Store undiluted at 4°C
For Use With (Application) Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry)
Dye Type DNA dye
Quantity 200 μL
Reactivity Tested in Development: Human, Mouse; Reported Reactivity: Rat
Excitation Wavelength 598/646 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.