missing translation for 'onlineSavingsMsg'
Learn More

BD Human Neural Cell Sorting Kit

Product Code. 15851769
Click to view available options
Quantity:
10 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15851769

Brand: BD 562271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • PE Mouse Anti-Human CD24
  • PerCP-Cy5.5 Mouse Anti-Human CD271
  • PerCP-Cy5.5 Mouse Anti-Human CD44
  • PE-Cy7 Mouse Anti-Human CD15
  • APC Mouse Anti-Human CD184
  • PE Mouse IgG1, k Isotype Control
  • PE-Cy7 Mouse IgM, k Isotype Control
  • APC Mouse IgG1, k Isotype Control
  • Negative Control CompBead Plus
  • Anti-Mouse Ig, k CompBead Plus
  • PerCP-Cy5.5 Mouse IgG1, k Isotype Control

Designed to allow the isolation of neural stem cells (NSCs) derived from human pluripotent stem cells or the isolation of neurons and glial cells from differentiated NSCs

This method greatly reduces the amount of variability between NSC isolations as opposed to traditional methods like manual isolation. In addition, four of the five included conjugates can be used to isolate neurons from glia from differentiated NSCs.

  • NSCs are sorted from the heterogeneous cell culture utilizing the included antibody conjugates to five cell surface marker

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human
Quantity 10 Tests
For Use With (Application) Designed to allow the isolation of neural stem cells (NSCs) derived from Human pluripotent stem cells or the isolation of neurons and glial cells from differentiated NSCs
Content And Storage 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.