missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Fusion Protein Buffer Kit
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
Description
- For use with Fusion Protein Kit
- 100 tests
Protein isolation and purification
Specifications
Specifications
| Buffer | Fusion Protein Buffer |
| Product Type | Fusion Protein Buffer kit |
| Quantity | 100 Tests |
| Includes | Pretreatment A (powder), pretreatment B (powder), capture beads, detecto r reagent |
| Content And Storage | Pretreatment A and B at -20°C, beads and reagent at 2° to 8°C |
| For Use With (Application) | Protein isolation and purification |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction