missing translation for 'onlineSavingsMsg'
Learn More

BD Caspase-3, Active Form, mAb Apoptosis Kit : FITC (CPP32)

Product Code. 15886278
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15886278

Brand: BD 550480

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

FITC-conjugated Monoclonal Rabbit Anti-Active Caspase-3 Antibody, BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution, and BD Perm/Wash™ Buffer

For Intracellular staining

  • Format: FITC
  • Clone: C92-605
  • Isotype: Rab IgG
  • Reactivity: Ms, Hu

Intracellular staining, Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage 4°C
For Use With (Application) Intracellular Staining
Conjugate FITC
Form Liquid

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.