Learn More
BD Caspase-3, Active Form, mAb Apoptosis Kit
Description
Includes
PE-conjugated Monoclonal Rabbit Anti-Active Caspase-3 Antibody, BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution, and BD Perm/Wash™ Buffer
Cysteine proteases plays key role in apoptosis and inflammation. Caspase-3 being key protease is activated during early stages of apoptosis and, like other members of caspase family, is synthesized as inactive pro-enzyme that is processed in cells undergoing apoptosis by self-proteolysis and/or cleavage by another protease. The processed forms of caspases consist of large (17-22 kDa) and small (10-12 kDa) subunits which associate to form active enzyme. Active caspase-3, a marker for cells undergoing apoptosis, consists of heterodimer of 17 and 12 kDa subunits which is derived from the 32 kDa pro-enzyme. Active caspase-3 proteolytically cleaves and activates other caspases, as well as relevant targets in cytoplasm and nucleus. This antibody has been reported to specifically recognize the active form of caspase-3 in human and mouse cells.
- Clone: C92-605
- Format: PE
- Isotype: IgG
Intracellular staining (flow cytometry)
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | 4°C |
| For Use With (Application) | Intracellular Staining |
| Form | Liquid |
Research use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.