missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II, I Plate™ Prepared Media

Product Code. 17382321
Click to view available options
Quantity:
100/Pk.
20/Pk.
Media Type:
Trypticase™ Soy Agar (TSA II), MacConkey II Agar
Unit Size:
Pack of 100
Pack of 20
This item is not returnable. View return policy

Product Code. 17382321

Brand: BD 221290

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Tripticase Soy Agar (TSA II)

  • For the visualization of hemolytic reactions produced by many bacterial species
MacConkey II Agar
  • Selective and differential plating medium for the detection of coliform organisms and enteric pathogens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar (TSA II), MacConkey II Agar
Formulation Trypticase™ Soy Agar with 5% Sheep Blood (TSA II): Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% ; MacConkey II Agar: Pancreatic Digest of Gelatin 17.0g/L, Sodium Chloride 5.0g/L, Pancreatic Digest of Casein 1.5g/L, Neutral Red 0.03g/L, Peptic Digest of Animal Tissue 1.5g/L,Crystal Violet 0.001g/L, Lactose 10.0g/L, Agar13.5g/L, Bile Salts 1.5g/L
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Packaging Type Plate
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.