missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ TSI Agar™ Slants (Triple Sugar Iron Agar) Prepared Media

Product Code. 17392311
Click to view available options
Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
This item is not returnable. View return policy

Product Code. 17392311

Brand: BD 221039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type TSI Agar (Triple Sugar Iron)
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Sodium Chloride 5g/L, Lactose 10g/L, Sucrose 10g/L, Dextrose 1g/L, Ferrous Ammonium Sulfate 0.2g/L, Sodium Thiosulfate 0.2g/L, Phenol red 0.025g/L, Agar 13g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Packaging Type Size K tubes with tight cap
Quantity 100/Pk.

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.