missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin

Product Code. p-4638890
Click to view available options
Quantity:
10 x 8 mL
100 x 5 mL
100 x 8 mL
Color:
Beige
Light Yellow
Packaging Type:
Size K tube with tight cap
Unit Size:
Pack of 10
Pack of 100
This item is not returnable. View return policy

Product Code. 17302351

Brand: BD 221742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CLSI, CMPH2, MCM9
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type Size K tube with tight cap
Quantity 100 x 5 mL

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.