missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Mycosel™ Prepared Agar Slants
Click to view available options
Quantity:
10/Pk.
Media Type:
Mycosel Agar
Unit Size:
Pack of 10
Description
- Selective medium containing cycloheximide and chloramphenicol
- For recovery of dermatophytes
Tubes, bottles and flasks with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, BS12, CMPH2, MCM9 |
| Color | Light Yellow |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Mycosel Agar |
| Formulation | Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L |
| For Use With (Application) | Isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria |
| Packaging Type | Tube |
| Quantity | 10/Pk. |
For in vitro Diagnostic Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction