missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Apoptotic Jurkat Lysate Set I
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
Includes
Jurkat Control Lysate and Camptothecin-treated Jurkat Lysate
- Used as positive control
- Cells in log-phase growth are induced for apoptosis
Western blotting
Specifications
Specifications
| Quantity | 1 Kit |
| Sample Volume | 500 μg |
| Storage Buffer | SDS-PAGE Buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% Bromophenol blue, 5% Glycerol) |
| Content And Storage | -20°C |
| Form | Liquid |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction