missing translation for 'onlineSavingsMsg'
Learn More

BD Apoptosis Detection Kit (APO-Direct)

Product Code. 15858678 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858678 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15858678 Supplier BD Supplier No. 556381
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

FITC-dUTP, PI/RNase Staining Buffer, Reaction Buffer, Rinsing Buffer, Wash Buffer, TdT Enzyme, Negative Control Cells, Positive Control Cells

Single-step staining method for labeling DNA breaks to detect apoptotic cells by flow cytometry.

  • Reagents are provided to process 50 tests

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
For Use With (Application) Flow Cytometry
Form Liquid
compliance-icons compliance-icons
Product Identifier
  • BD Pharmingen APO-DIRECT Kit
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: arsenic compounds, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.