missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9-2 Antibody (1C9G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | BCL9-2 |
|---|---|
| Clone | 1C9G2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18255131
|
Novus Biologicals
NBP2-61705 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631969
|
Novus Biologicals
NBP2-61705-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL9-2 Monoclonal specifically detects BCL9-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| BCL9-2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, BCL9-like protein, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2 | |
| BCL9L | |
| IgG1 | |
| Protein G purified |
| 1C9G2 | |
| Western Blot 1:5000-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| 283149 | |
| Purified recombinant fragment of human BCL9L (AA: 606-751) expressed in E. Coli. | |
| Primary | |
| 157 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title