missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9-2 Antibody (1C9G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61705-0.025ml
This item is not returnable.
View return policy
Description
BCL9-2 Monoclonal specifically detects BCL9-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| BCL9-2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, BCL9-like protein, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2 | |
| Mouse | |
| 157 kDa | |
| 0.025 mL | |
| 283149 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 1C9G2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BCL9L | |
| Purified recombinant fragment of human BCL9L (AA: 606-751) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction