missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£184.00 - £260.00
Specifications
| Antigen | Bcl-xL |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084955
|
Novus Biologicals
NB100-56104SS |
0.025 mL |
£184.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18096454
|
Novus Biologicals
NB100-56104 |
0.05 mL |
£260.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-xL Polyclonal specifically detects Bcl-xL in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Bcl-xL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Whole antisera with 0.05% Sodium Azide | |
| 598 | |
| A synthetic peptide corresponding to amino acids 46-66 (ETPSAINGNPSWHLADSPAVN) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 100% conserved in Bcl-XS. The immunogen sequence is 100% in multiple species including mouse, rat, dog, cow, gerbil, and sheep. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| BCL2L1 | |
| IgG | |
| Unpurified | |
| Bcl-XL+S |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title