missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56104
This item is not returnable.
View return policy
Description
Bcl-xL Polyclonal specifically detects Bcl-xL in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Bcl-xL | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Bcl-XL+S | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| BCL2L1 | |
| A synthetic peptide corresponding to amino acids 46-66 (ETPSAINGNPSWHLADSPAVN) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 100% conserved in Bcl-XS. The immunogen sequence is 100% in multiple species including mouse, rat, dog, cow, gerbil, and sheep. | |
| 0.05 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 598 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction