missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Antibody (5E9), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | bcl-x |
|---|---|
| Clone | 5E9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30033014
|
Novus Biologicals
NBP3-26480-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025849
|
Novus Biologicals
NBP3-26480-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| bcl-x | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Apoptosis regulator Bcl-X,BCL2L,BCL2L1,Bcl-2-like protein 1,BCLX,Bcl-X,BCL-XL/S,PPP1R52 | |
| A synthesized peptide derived from Human Bcl-x [UniProt Q07817] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5E9 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 598 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title