missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Antibody (5E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26480-50ul
This item is not returnable.
View return policy
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| bcl-x | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 5E9 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Apoptosis regulator Bcl-X,BCL2L,BCL2L1,Bcl-2-like protein 1,BCLX,Bcl-X,BCL-XL/S,PPP1R52 | |
| A synthesized peptide derived from Human Bcl-x [UniProt Q07817] | |
| 50 μL | |
| Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
| 598 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction