Learn More
Epredia™ Lab Vision™ Bcl-6 Ab-2, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Bcl-6 Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1114-RQ
Code : LD
Additional Details : Weight : 0.01200kg
Description
Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.Host Species: Mouse
Clone: BL6.02, same as PG-B6p
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Rabbit, Rat, Sheep and Pig. Others not tested.
Epitope: aa 3-484
Immunogen: Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Bcl-6 Ab-2 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein | |
RUO | |
Primary |
Immunohistochemistry (Paraffin) | |
BL6.02; same as PG-B6p | |
Mouse | |
12 mL | |
Apoptosis | |
Ready-to-Use |