missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Bcl-6 Ab-2, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Bcl-6 Ab-2, Mouse Monoclonal Antibody.
£113.22 - £535.00
Specifications
Antigen | Bcl-6 Ab-2 |
---|---|
Clone | BL6.02; same as PG-B6p |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12609016
|
Epredia™
MS-1114-S1 |
Concentrated | 500 μL |
£348.00
500µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12689006
|
Epredia™
MS-1114-S |
Concentrated | 1 mL |
£535.00
1mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12679006
|
Epredia™
MS-1114-RQ |
Ready-to-Use | 12 mL |
£510.00
12mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12669006
|
Epredia™
MS-1114-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12699006
|
Epredia™
MS-1114-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.Host Species: Mouse
Clone: BL6.02, same as PG-B6p
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Rabbit, Rat, Sheep and Pig. Others not tested.
Epitope: aa 3-484
Immunogen: Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Bcl-6 Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein |
BL6.02; same as PG-B6p | |
Monoclonal | |
Mouse | |
Apoptosis | |
Primary |