missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£168.00 - £248.00
Specifications
| Antigen | Bcl-2 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099583
|
Novus Biologicals
NB100-56102SS |
0.025 mL |
£168.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18090826
|
Novus Biologicals
NB100-56102 |
0.05 mL |
£248.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-2 Polyclonal specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Bcl-2 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Tumor Suppressors | |
| Whole antisera with 0.05% Sodium Azide | |
| 596 | |
| A synthetic peptide corresponding to amino acids 61-76 (ASRDPVARTSPLQTPAC) of human Bcl-2 was used as immunogen; GenBank no NP_000648.2. This sequence is 100% conserved in human Bcl-2 alpha (239 amino acids) and beta (205 amino acids) isoforms. The literature indicates that the immunogen sequence is not conserved between human Bcl-2 and other species. Therefore this antibody may be particularly useful for detecting human Bcl-2 in transgenic studies. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
| BCL2 | |
| IgG | |
| Unpurified | |
| Bcl-2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title