missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 Antibody, Novus Biologicals™

Product Code. 18099583 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
Unit Size:
0.05mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18099583 0.025 mL 25µL
18090826 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18099583 Supplier Novus Biologicals Supplier No. NB10056102SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

Bcl-2 Polyclonal specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Gene Symbols BCL2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 61-76 (ASRDPVARTSPLQTPAC) of human Bcl-2 was used as immunogen; GenBank no NP_000648.2. This sequence is 100% conserved in human Bcl-2 alpha (239 amino acids) and beta (205 amino acids) isoforms. The literature indicates that the immunogen sequence is not conserved between human Bcl-2 and other species. Therefore this antibody may be particularly useful for detecting human Bcl-2 in transgenic studies.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 596
Test Specificity Bcl-2
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.