missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Antibody (bcl-2/124) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £319.00
Specifications
| Antigen | Bcl-2 |
|---|---|
| Clone | bcl-2/124 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605728
|
Novus Biologicals
NBP2-50116-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673947
|
Novus Biologicals
NBP2-50116 |
0.1 mL |
£319.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Bcl-2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Autophagy, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Oncogenes, Tumor Suppressors | |
| PBS | |
| 596 | |
| IgG1 | |
| Protein A purified |
| bcl-2/124 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
| Synthetic peptide of amino acids 41-54 of human Bcl-2 oncoprotein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title