missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 Antibody (bcl-2/124) - BSA Free, Novus Biologicals™

Product Code. 18605728 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18605728 0.025 mL 0.03mL
18673947 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18605728 Supplier Novus Biologicals Supplier No. NBP2501160.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone bcl-2/124
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Host Species Mouse
Immunogen Synthetic peptide of amino acids 41-54 of human Bcl-2 oncoprotein
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Oncogenes, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.