missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57574
This item is not returnable.
View return policy
Description
BCKDK Polyclonal specifically detects BCKDK in Human samples. It is validated for Western Blot.
Specifications
| BCKDK | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BCKDHKIN, branched chain alpha-ketoacid dehydrogenase kinase, branched chain ketoacid dehydrogenase kinase, EC 2.7.11, EC 2.7.11.4, mitochondrial | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Pig: 100%; Canine: 92%; Equine: 91%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O14874 | |
| BCKDK | |
| Synthetic peptides corresponding to BCKDK(branched chain ketoacid dehydrogenase kinase) The peptide sequence was selected from the N terminal of BCKDK. Peptide sequence CLPFIIGCNPTILHVHELYIRAFQKLTDFPPIKDQADEAQYCQLVRQLLD. | |
| 100 μL | |
| Protein Kinase | |
| 10295 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction