missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58142
This item is not returnable.
View return policy
Description
BCAT1 Polyclonal specifically detects BCAT1 in Human samples. It is validated for Western Blot.
Specifications
| BCAT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Rabbit: 100%; Equine: 92%; Chicken: 91%; Sheep: 91%; Mouse: 84%. | |
| Human, Mouse, Pig, Canine, Equine, Rabbit, Sheep | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P54687 | |
| BCAT1 | |
| Synthetic peptides corresponding to BCAT1(branched chain aminotransferase 1, cytosolic) The peptide sequence was selected from the N terminal of BCAT1. Peptide sequence MKDCSNGCSAECTGEGGSKEVVGTFKAKDLIVTPATILKEKPDPNNLVFG. | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 586 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction