missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC Plates

Product Code. p-4770203
Click to view available options
Quantity:
100/Pk.
Color:
Yellow
Media Type:
Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack
This item is not returnable. View return policy

Product Code. 16338951

Brand: BD 222207

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Rodac (Replicate Organism Detection and Counting) plates for surface sampling.

To use:

  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Color Yellow
Content And Storage 2°C to 8°C
Format Rodac
Manufacturing Origin United States
Media Type Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack
Sterility Sterile
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Monitoring surfaces in clean rooms and other environmentally controlled areas
Packaging Type Gamma-irradiated Double Wrap
Quantity 100/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.