missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC Plates
Click to view available options
Quantity:
100/Pk.
Color:
Yellow
Media Type:
Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack
Description
Rodac (Replicate Organism Detection and Counting) plates for surface sampling.
To use:
- Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
- When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
- Use the included inner bag to transport plates from your critical environment
Compliance
Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Color | Yellow |
| Content And Storage | 2°C to 8°C |
| Format | Rodac |
| Manufacturing Origin | United States |
| Media Type | Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack |
| Sterility | Sterile |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L |
| For Use With (Application) | Monitoring surfaces in clean rooms and other environmentally controlled areas |
| Packaging Type | Gamma-irradiated Double Wrap |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction