missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Mycosel™ Prepared Agar Slants

Product Code. p-4639617
Click to view available options
Quantity:
10/Pk.
Media Type:
Mycosel Agar
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17312311

Brand: BD 220966

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For the isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria

  • Selective medium containing cycloheximide and chloramphenicol
  • For recovery of dermatophytes

Tubes, bottles and flasks with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, MCM9
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Mycosel Agar
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L
For Use With (Application) Isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria
Packaging Type Tube
Quantity 10/Pk.

For in vitro Diagnostic Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.