Learn More
RETSCH BB 300 Model Jaw Crushers
Designed for the rapid, effective crushing and pre-crushing of medium-hard, hard, brittle and tough materials. Retsch™ BB 300 Model Jaw Crushers are ideal for sample preparation in laboratories and industrial plants.
Brand:  RETSCH 20.057.1002
Additional Details : Weight : 672.00000kg
 View more versions of this product
Product Code. 10779312
  • £54160.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • Excellent crushing performance
  • Wear compensation with zero-point adjustment
  • Infinitely gap width setting
  • Overload protection
  • Wide range of materials for contamination free grinding
  • No-rebound feed hopper with quick-release clamp
  • Brake motor with safety switch
  • Easy-to-clean crushing chamber
  • Connector for dust extraction
  • Central lubrication
  • Suitable for integration in automatic installations
  • Special version for grinding semi-conductor materials
Specifications
Specifications
Jaw Crusher
1600 x 670 x 1450 mm
1450 mm
1600 mm
BB 300, For on-site integration
670 mm
CE Certified
Chemistry/Plastics, construction materials, engineering/electronics, environment/recycling, geology/metallurgy, glass/ceramics
IP54
Stainless Steel
3-phase Power Connection